This item is now discontinued. Please browse similar items
Marathon Septa, Not Pre-Pierced, Shimadzu; 50/Pack
Representative image only.
Marathon Mfr # 239498 – Item # RK-01999-08
Long injection life, pre-pierced for minimal coring
  • CenterGuide™ design requires less force for initial penetration
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithShimadzu
  • DescriptionSepta, Not Pre-Pierced, Shimadzu; 50/Pack
  • Warranty1 month
1 Month Warranty
Key Features
  • CenterGuide™ design requires less force for initial penetration
  • 1 month warranty
More About this Item
Marathon™ Septa have outstanding durability in either autosampler or manual injection applications. Operate at a maximum temperature of 400°C and achieve up to 400 injections per septum. Pre-piercing minimizes injection coring on select CenterGuide septa.