MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 9, 1 pair
Representative image only.
MAPA Mfr # 65031-9 – Item # RK-86357-42
Inner fabric lining provides exceptional comfort
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
PRODUCT OPTIONS
$114.19CAD / Pair of 1
BUY MORE & SAVE
  • 1-5$114.19CAD / Pair of 1
  • 6+$102.77CAD / Pair of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialButyl
  • Thickness (mil)60
  • Size9
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • LiningCotton
  • CuffScalloped
  • SterileNo
  • DescriptionCotton-Lined Butyl Gloves, size 9, 1 pair
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Butyl gloves provide excellent chemical resistance in applications involving acids, ketones, esters, and amine derivatives. 100% seamless cotton liner provides extra comfort for long-term use. Scalloped cuffs can be rolled down to prevent drips from reaching your arms. Unique Z-pattern grip design for optimal grip.