This item is now discontinued. Please browse similar items
MAPA 333319 Heat-Insulated Nitrile Gloves, size 9
Representative image only.
MAPA Mfr # 333319 – Item # RK-40211-80
“All-in-one” thermal and liquid protection
  • Two special thermal liners protect workers from hot or cold temperature
  • Middle nitrile polymer layer keeps hands dry in wet or chemical applications
  • Provides maximum thermal insulation from -22 to 480°F (-30 to 249°C)
  • Black color hides dirt, exten
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Size9
  • Length (in)12
  • Length (cm)30.5
  • ColorBlack
  • LiningThermal
  • SterileNo
  • DescriptionHeat-Insulated Nitrile Gloves, size 9
  • Warranty1 year
1 Year Warranty
Key Features
  • Two special thermal liners protect workers from hot or cold temperature
  • Middle nitrile polymer layer keeps hands dry in wet or chemical applications
  • Provides maximum thermal insulation from -22 to 480°F (-30 to 249°C)
  • Black color hides dirt, exten
  • 1 year warranty
More About this Item
These gloves feature two special thermal liners to protect workers from hot or cold temperatures, with a nitrile polymer layer in the middle to keep hands dry in wet or chemical applications. Provide maximum thermal insulation from -22 to 480°F (-30 to 249°C). Black color hides dirt. Gloves are launderable to reduce costs and environmental waste.