Low-cost, Direct-drive Air Motor Mixer; 200-10,000 RPM
Representative image only.
Mfr # TYPE A – Item # RK-04680-00
  • 1 year warranty
PRODUCT OPTIONS
$856.04CAD / Each
$371.29CAD / Each
for an additional
$371.29CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Speed (rpm)200
  • Max Speed (rpm)10000
  • Max Torque (in-oz)64
  • Motor hp1/3
  • DescriptionLow-cost, Direct-drive Air Motor Mixer; 200-10,000 RPM
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Safely use these air-drive mixers whenever electrical power is hazardous or impractical. Ideal for mixing solvents, lacquers, and volatile chemicals without the possibility of spark ignition.
Mixers accept an air input line pressure to 80 or 100 psi (10 to 12 cfm). A needle valve enables you to control mixing speed throughout the range for each mixer, and a built-in muffler ensures quiet operation. We recommend installing an air filter/regulator 07042-75 and an air lubricator 07042-70 in your air supply line.
Includes: 316 SS paddle assembly (3⁄8" dia x 12"L [10 mm dia x 305 mm L] shaft with 21⁄2" [64 mm] dia propeller), collet, 10"L x 9⁄16" dia (254 mm L x 14 mm dia) mounting rod, muffler, needle valve, and 4-ft (1.2-m) L air hose with fittings.