This item is now discontinued. Please browse similar items
Lovibond AF650 Petro Benchtop Petroleum Comparator
Representative image only.
Lovibond Mfr # 345030 – Item # RK-99562-36
Economical, visual color measurement with three-field determination
  • Available with the most popular color scales
  • Quick comparison of sample to glass color standards
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • TestPetroleum, oils, waxes
  • DescriptionBenchtop Petroleum Comparator
  • Warranty1 year
1 Year Warranty
Key Features
  • Available with the most popular color scales
  • Quick comparison of sample to glass color standards
  • 1 year warranty
More About this Item
Convenient benchtop comparator is a three-field instrument for visually determining the color of samples by direct comparison with colored glass filters. With a 3-section field of view, the sample and two consecutive glasses on the color scale are viewed simultaneously, making it easier to achieve the optimal color match. For rapid color grading within predetermined color limits, the glasses can be set to the two limiting colors, making it easy to see if the sample is within tolerance.