This item is now discontinued. Please browse similar items
Labomed 4145000 Stereozoom Camera Microscope
Representative image only.
Labomed Mfr # 4145000 – Item # RK-48923-90
Built-in 5-megapixel camera captures images and live video
  • Adjustable-angle top light helps reduce glare from reflective items
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification8x to 35x
  • Maximum Magnification35x
  • Head ConfigurationBinocular
  • Resolution5 MP
  • Illuminator(s)LED, 10-bulb array on top, 3-bulb array on bottom
  • Image Sensor1/2.5\" CMOS
  • Cable(s)USB, NTSC/PAL
  • Power (VAC)100 to 240
  • DescriptionStereozoom Camera Microscope
  • Warranty5 Years
5 Year Warranty
Key Features
  • Adjustable-angle top light helps reduce glare from reflective items
  • 5 year warranty
More About this Item
This microscope offers ergonomic zoom knob and focus controls to make use more comfortable. Metal gear system is durable with smooth control. The built-in 5 megapixel camera provides high-resolution images or video of your sample. Variable-intensity LED illumination provides excellent lighting and long service life. Top illuminator has three settings: illuminate all ten LEDs, the front five LEDs, or the back five LEDs. You can also add an oblique ±30° angle to the top illuminator to cut down on glare from reflective items.
Additional features include removable Abbe condenser, diopter adjustment, and USB output.