This item is now discontinued. Please browse similar items
Labnet D1296-PCR Dual block, 96 well PCR plate, skirted or nonskirted, for dual block unit only
Representative image only.
Labnet Mfr # D1296-PCR – Item # RK-37885-17
Uniformly molded high-grade aluminum blocks provide excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Heaters sold separately
  • PopStopper and AccuRack accessory sold seperately
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")6 1/100
  • Width (InchShort)3 35/64
  • Uniformity (° F)0.36
  • Uniformity (° C)0.2
  • Block Number Of Openings96
  • DescriptionLabnet AccuBlock D1296-PCR Dual block, 96 well PCR plate, skirted or nonskirted, for dual block unit only
  • Warranty1 year
1 Year Warranty
Key Features
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Heaters sold separately
  • PopStopper and AccuRack accessory sold seperately
  • 1 year warranty
More About this Item
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Thermometer wells available in all blocks except those used with plates.