Labmat F246750000 Liner, 20" W; 50 ft/Roll, 1 Roll/Pk

Representative image only.
Labmat Mfr # F246750000 – Item # RK-06805-01
Skid resistent polyethylene film backing that prevents leakage
  • Disposable

PRODUCT OPTIONS

$132.31CAD / Roll of 1

SPECS & DESCRIPTION

Specifications & Description

  • Length (\")600
  • MaterialPolyethylene
  • Width (InchShort)20
  • Width (in)20
  • Length (ft)50
  • Length (in)600
  • Width (cm)50.8
  • Length (m)15.24
  • Length (cm)1524
  • ColorWhite
  • DescriptionLiner, 20\" W; 50 ft/Roll, 1 Roll/Pk

Key Features

  • Disposable

More About this Item

Highly abosrbent top layer soaks up corrosive materials, and most liquids. Red backing alerts end-users when biohazard materials are in use. Liners can easily be cut to meet end users specific applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat