This item is now discontinued. Please browse similar items

Labconco RXPert Tall & Deep Double Filtered Balance System, No Surface, 5', 115 V

Representative image only.
Labconco Mfr # 3971590 – Item # RK-32521-67
Keep hazardous powders and particulates contained
  • Meet USP <800> requirements for nonsterile hazardous compounding with redundant filtration optional
  • Bag in/bag out HEPA filter provides protection during filter changes
  • Outside ducting optional
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)60
  • Exterior Width (in)60
  • Exterior Height (in)58.8125
  • Exterior Depth (in)35.5
  • Interior Width (in)57.625
  • Interior Height (in)32
  • Interior Depth (in)30
  • Power (VAC)115
  • Power (Hz)60
  • Sash opening (in)12
  • Exhaust Volume (CFM)480
  • Shipping Weight (lbs)745
  • Shipping Weight (kg)338
  • DescriptionTall & Deep Double Filtered Balance System, No Surface, 5', 115 V
  • Warranty1 Year
1 Year Warranty

Key Features

  • Meet USP <800> requirements for nonsterile hazardous compounding with redundant filtration optional
  • Bag in/bag out HEPA filter provides protection during filter changes
  • Outside ducting optional
  • 1 year warranty

More About this Item

The RXPert cabinet features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement. The filtered balance system is a Class I enclosure that complies with USP requirements for nonsterile compounding.


All models include a built-in analog airflow monitor. Some models include a stainless steel or epoxy work surface; a work surface is required if one is not included.


Double-Filtered Enclosures feature redundant filters in series, so do not require exhausting to the outside while maintaining USP <800> compliance. All models require a supporting base (order separately).


Compliance: USP <800>, SEFA 1-2010, modified ASHRAE 110-1995, ANSI Z9.5-2011, UL 61010-1, and CAN/CSA C22.2 No. 61010.1 standards.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat