This item is now discontinued. Please browse similar items

Labconco Purifier 4 ft Horizontal Clean Bench, 39" D/115V

Representative image only.
Labconco Mfr # 3888401 – Item # RK-09116-12
Remove particles from room air to supply Class 100 air to your bench working area
  • Provides ISO Class 5 conditions and product protection
  • Two interior depths offered to match your application needs
  • Easy to clean, static-dissipative stainless steel work surface
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)48
  • Blower TypeStandard
  • Exterior Width (in)50
  • Exterior Height (in)48.4375
  • Exterior Depth (in)39
  • Power (VAC)115
  • Power (Hz)60
  • Shipping Weight (lbs)465
  • Shipping Weight (kg)211
  • Description4 ft Horizontal Clean Bench, 39\" D/115V
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneETL Listed

Key Features

  • Provides ISO Class 5 conditions and product protection
  • Two interior depths offered to match your application needs
  • Easy to clean, static-dissipative stainless steel work surface
  • 1 year warranty

More About this Item

Sterile horizontal laminar flow benches filter out particles as small as 0.3µm with 99.99% efficiency, providing ISO Class 5 air (fewer than 3520 particles, 0.5 µm or larger per cubic meter of air). Prefilters trap large particles, extending HEPA filter life. Differential pressure gauge displays pressure across the HEPA filter.

Benches are constructed of powder-coated steel exterior, safety glass side panels with utility ports, and 304 stainless steel work surface. Models with built-in 254 nm UV light feature retractable UV screen. Light timer provides timed UV light exposure (5, 10, 15, 30, 60, 120, 240 minutes and continuous settings). Benches include a built-in motor blower with vibration isolation pad. Other 230-volt plug types are available; contact an Application Specialist for details.

Note: Benches provide sample protection only. No personnel protection is provided. Do not use with biohazards, toxins or radionuclides.

Compliance: ETL (115 volt models), CE (230 volt models)

Accessories

Labconco Telescoping Base Stands — support Purifier® Horizontal Clean Benches

Stands feature telescoping legs so you can select stand height during installation. Height is adjustable to eight positions in 1” increments from 27.5” to 34.5”. Frame is durable 1.75” tubular, powder-coated steel with rear crossbar supports. ADA compliant. Fixed leveling feet.

Labconco Replacement Filters for Purifier® Horizontal Clean Benches

*38-39” W and 50-51” W clean benches require two prefilters, 74-75” wide clean benches require three prefilters, and 99-100” wide clean benches require four prefilters.
** 99-100” wide clean benches require two HEPA filters.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat