This item is now discontinued. Please browse similar items
Labconco Echo Filtered Fume Hood Work Surface 8', No Cupsink Cutouts
Representative image only.
Labconco Mfr # 9501800 – Item # RK-32535-54
  • Supports all Labconco benchtop fume hoods (work surface is required).
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Hood Width (in)96
  • DescriptionFiltered Fume Hood Work Surface 8', No Cupsink Cutouts
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Supports all Labconco benchtop fume hoods (work surface is required).
  • 1 year warranty
More About this Item
Work surfaces are solid epoxy resin and available flat or with SpillStopper surface, which have dished surfaces to keep accidental spills and liquids way from the seam between the hood wall and surrounding countertop. SpillStopper Surfaces support Protector Premier Hoods. Flat Surfaces support Fiberglass 30 and Basic 47 and 70 Hoods. They are offered without cutouts for a sink. Contact Cole-Parmer for work surfaces with pre-drilled cutouts.