Labconco 9015320 WaterPro BT Water Purification System, Remote Dispense, UV Lamp; 115V

Representative image only.
Labconco Mfr # 9015320 – Item # RK-99290-46
Compact benchtop design produces both Type I and Type III (RO) water!
  • Set up in minutes—simply connect to tap supply, insert the filter pack, and power on
  • Integrated 6-liter storage tank holds Type III RO-purified water
  • Quiet—50 dB—and compact enough to be shared between labs, set up on a benchtop, or wall mounted
  • Integrated UV lamp (185 and 254 nm) removes bacteria, their metabolites, and certain organics
  • Type I water is dispensed from the Remote Dispenser, with the option of Auto-Dispense mode for preset volumes
  • Remote dispenser allows the unit to be placed up to 6.5 ft (2 m) away from the dispensing location
  • 1 year warranty

PRODUCT OPTIONS

$12,420.68CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ASTM Water GradeType I and Type III-IV
  • Capacity10 L/day
  • Max Flow Rate (L/min)0.5
  • With UV LampYes
  • Conductivity Threshold< 2000 microseimens/cm
  • SourceTap water
  • TOC Threshold< 2000 ppb
  • Feed Water Requirements (Source)Tap water
  • Type I Water (TOC)<5 ppb
  • Type I Water (Bacteria)< 01 cfu/mL
  • Max Inlet Water Temperature (° F)95
  • Min Inlet Water Temperature (° F)41
  • Max Inlet Water Temperature (° C)35
  • Min Inlet Water Temperature (° C)5
  • Min Inlet Pressure (PSI)7.3
  • Max Inlet Pressure (PSI)87
  • Min Inlet Pressure (bar)0.5
  • Max Inlet Pressure (bar)6
  • Power (VAC)115
  • DescriptionWaterPro BT Water Purification System, Remote Dispense, UV Lamp; 115V
  • Warranty1 year
1 Year WarrantyUL Listed to Canadian safety standards.Conformite EuropeenneCanadian Standards AssociationUL Listed.

Key Features

  • Set up in minutes—simply connect to tap supply, insert the filter pack, and power on
  • Integrated 6-liter storage tank holds Type III RO-purified water
  • Quiet—50 dB—and compact enough to be shared between labs, set up on a benchtop, or wall mounted
  • Integrated UV lamp (185 and 254 nm) removes bacteria, their metabolites, and certain organics
  • Type I water is dispensed from the Remote Dispenser, with the option of Auto-Dispense mode for preset volumes
  • Remote dispenser allows the unit to be placed up to 6.5 ft (2 m) away from the dispensing location
  • 1 year warranty

More About this Item

WaterPro BT systems deliver up to 10 L/day of ASTM Type I water direct from a tap feed water supply. Ideal for lab applications such as trace elemental analysis, mixing or diluting acid/base solutions, and media preparation.

In addition, ASTM Type III RO-purified water is produced at a rate of 3 L/hr* and stored in an integrated 6-L tank. Ready and waiting for glassware rinsing, reagent preparation, and other noncritical laboratory water needs.

Compact, lightweight, and simple to install, these systems can be easily transported and shared among labs. All water filtration technologies-activated carbon filtration, deionization, and RO-are combined into one convenient twin-cartridge pack that snaps into place (order required filter pack 99290-48 separately).

User-friendly color LCD shows water quality, system status, and maintenance or alarm messages. Plus, it can be tilted for easy viewing from any angle, even if system is wall mounted.

Keypad allows quick access to display settings, water quality unit of measure, resistivity set point, and temperature values. It also provides a means to set the Type I water volume to be dispensed, dispensing time, flow rate calibration, and tank level calibration.

For further purification, choose from two optional final filters that attach to the dispensing valve. The hollow fiber filter removes all particles and bacteria larger than 0.22 micron. The ultrafilter cartridge removes pyrogens (pathogens, RNase, and DNase).

*Feedwater conductivity must be 100 µS or better.

Remote Dispense option adds the remote dispenser with 6.5-ft (2-m) tether and powder-coated steel dispenser stand, and the UV lamp option also includes the lamp needed for oxidizing purified water at 185- and 254-nm.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat