This item is now discontinued. Please browse similar items
Labconco 7910013 RapidVap N2/48 Evaporation Systems, 230 V
Representative image only.
Labconco Mfr # 7910013 – Item # RK-26853-36
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)230
  • Power (Hz)50
  • DescriptionRapidVap N2/48 Evaporation Systems, 230 V
  • Warranty1 year
1 Year WarrantyETL Listed
Key Features
  • 1 year warranty
More About this Item
N2 Evaporation System offers a more efficient, automatic alternative to Kuderna-Danish evaporators. Vortex mixing maximizes solvent surface area for efficient evaporation. Nitrogen is directed onto the sample surface to reduce solvent partial-pressure and speed evaporation. 1000-watt heater distributes heat evenly to all vessels through PTFE®-coated aluminum blocks for rapid yet gentle drying. Two heat-zone design enables quick evaporation while protecting concentrated sample from going to dryness.
Vortex maximizes sample surface area for evaporation while minimizing bumping and sample loss. Vortex mixing promotes homogeneity and minimizes physical damage to solids throughout the evaporation process. Reconstitution or re-suspension can be easily accomplish