This item is now discontinued. Please browse similar items
Labconco 5065300 Protector Glove Box Carbon Exhaust Filter
Representative image only.
Labconco Mfr # 5065300 – Item # RK-34760-51
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionProtector Glove Box Carbon Exhaust Filter
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Carbon Exhaust Filter is required for removal of gaseous organic contaminants in radioisotope or chemical carcinogen applications.