This item is now discontinued. Please browse similar items
Jeio Tech Lab Companion AAA44523 Universal Platform For 37l Shaking Bath
Representative image only.
Jeio Tech Lab Companion Mfr # AAA44523 – Item # RK-12054-44
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionUniversal Platform For 37l Shaking Bath
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Change your shaker's capacities with convenient parts and accessories.
Maximum flask capacities:
(20) 50mL flask clamps
(15) 100mL flask clamps
(8) 250mL flask clamps
(6) 500mL flask clamps
(4) 1000mL flask clamps
(2) 2000mL flask clamps