This item is now discontinued. Please browse similar items
Koehler K41500-4 Fluorescent Indicator Apparatus Silica Gel; 500 gram
Representative image only.
Koehler Mfr # K41500-4 – Item # RK-59876-87
Optimize performance of your Koehler Fluorescent Indicator Adsorption Apparatus
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFluorescent Indicator Apparatus Silica Gel; 500 gram
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Compatible for use with the Koehler Fluorescent Indicator Absorption Apparatus, sold separately.
An integrated electric vibration system facilitates the dry gel packing of the columns.
After the sample is introduced and adsorbed into the column, fluorescent dye indicators mark the boundaries of aromatics, olefins, and saturates, which are visible with a UV lamp.