This item is now discontinued. Please browse similar items
Koehler K33810 Steam Superheater, 220-240 VAC, 50/60 Hz
Representative image only.
Koehler Mfr # K33810 – Item # RK-59876-14
Optimize performance of your Koehler Existent Gum Evaporation Bath
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)230
  • Power (Hz)50/60
  • DescriptionSteam Superheater, 220-240 VAC, 50/60 Hz
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Compatible for use with the Koehler Existent Gum Evaporation Bath model K33700. Bath sold separately.
Steam Superheater provides dry superheated steam for evaporation baths not equipped with a built-in superheater. Use together with an outside steam source for steam-jet method testing of aircraft turbine fuels. Superheating chamber and condensate trap are constructed entirely of stainless steel. Solid state temperature controller adjusts between 0-550°F. Equipped with steam inlet and outlet connections and condensate drain valve. Steel exterior has a chemical resistant polyurethane enamel finish.