This item is now discontinued. Please browse similar items
Koehler K23700 Constant-Temperature Viscosity Bath, 22 L, 115 VAC
Representative image only.
Koehler Mfr # K23700 – Item # RK-98944-00
Save time – seven viscometer holders and seven bath timers allow multiple measurements at the same time
  • Motorized stirrer provides uniform temperature control and thorough circulation without turbulence
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Bath TypeViscosity Water Bath
  • Reservoir Capacity (L)22
  • Max Temperature (° C)150
  • Min Temperature (° C)Ambient
  • Temperature Accuracy (° C)Exceeds ASTM requirements
  • Temperature Sensor100 Ω Pt RTD
  • InterfaceNone
  • Wetted MaterialsGlass and SS
  • Power (VAC)115
  • DescriptionConstant-Temperature Viscosity Bath, 22 L, 115 VAC
  • Warranty1 year
1 Year Warranty
Key Features
  • Motorized stirrer provides uniform temperature control and thorough circulation without turbulence
  • 1 year warranty
More About this Item
Baths feature PID microprocessor controller to deliver precise temperature control, with ASTM specified tolerances throughout the operating temperature range. Dual digital displays show set point time and actual temperature. Timers display in 0.01-second resolution and have ±0.01% accuracy.
Controller automatically displays test results in efflux time and viscosity units when timers stop.
Controller automatically shuts off power when there is an overtemperature condition or if the primary probe is disconnected. The integrated low-level sensor interrupts operations if the bath liquid does not fill to the proper level, and shuts off power if the liquid falls below the level.
Bath chamber is a clear Pyrex® tank enclosed in a polyester-epoxy finished steel housing. Top plate holds the viscometers and is made of stainless steel for easy cleaning. Front viewing window reduces distortion. Glare-free fluorescent lights in the bath and a background baffle enhance the view of the viscometers. Baths rest on adjustable leveling feet.
All models conform to ASTM D445, D2170, D6074, D6158; IP 71, 319; ISO 3104; DIN 51550; FTM 791-305.