This item is now discontinued. Please browse similar items

Knick STRATOS pH PH/ORP 2-wire explosion-proof controller

Representative image only.
Knick Mfr # A201B-MSPH/1 – Item # RK-56030-10
Unique color-guided backlit display guarantees maximum operating safety
  • FM approved and CSA Class 1. Div 2 rated
  • High-contrast display with unique color-guided backlighting; changes between six different colors to clearly signal operating states
  • User-friendly operating system with plain-text ticker line
  • Two current outputs
  • Digital and analog input
  • Event logbook up to 200 entries
  • HART® communication system
  • All the electronics are easily accessible in the door module
  • Wall, pipe, or panel mounting
  • Connection with Memosens® technology digital pH sensors
  • Sensor identification and diagnostic with remaining lifetime indicator
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Control TypeOn/Off
  • Display TypeLCD
  • Housing MaterialNEMA 4X
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1mV
  • Min Temperature (° C)20
  • Max Temperature (° C)200
  • Min Temperature (° F)68
  • Max Temperature (° F)392
  • Temperature Resolution0.1°
  • Temperature CompensationAutomatic
  • Password ProtectionYes
  • Output Current4-20mA
  • Battery4 to 20 mAloop powered (14 to 30 VDC)
  • Width (cm)14.76
  • Height (cm)14.76
  • Width (in)5 13/16
  • Height (in)5 13/16
  • Depth (in)5 5/8
  • DescriptionPH/ORP 2-wire explosion-proof controller
  • Warranty3 year
3 Year WarrantyConformite EuropeenneCanadian Standards AssociationFM ApprovedIntrinsically SafeIP 67

Key Features

  • FM approved and CSA Class 1. Div 2 rated
  • High-contrast display with unique color-guided backlighting; changes between six different colors to clearly signal operating states
  • User-friendly operating system with plain-text ticker line
  • Two current outputs
  • Digital and analog input
  • Event logbook up to 200 entries
  • HART® communication system
  • All the electronics are easily accessible in the door module
  • Wall, pipe, or panel mounting
  • Connection with Memosens® technology digital pH sensors
  • Sensor identification and diagnostic with remaining lifetime indicator
  • 3 year warranty

More About this Item

These units offer Zone 2 intrinsic safety rating (FM approved and CSA Class 1. Div 2 rated) for use in hazardous areas. They accept Memosens® digital electrodes for additional value; using precalibrated sensors reduces costly onsite handling time and increases plant availability.

2-wire analyzer is fully functional with 2-wire supply and has an RS-485 interface for operating the Memosens sensors.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat