This item is now discontinued. Please browse similar items
Kinetronics ASC Anti-Static Cloth, synthetic and conductive fiber mesh
Representative image only.
Kinetronics Mfr # ASC – Item # RK-46301-00
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAnti-Static Cloth, synthetic and conductive fiber mesh
  • Warranty1 month
1 Month Warranty
Key Features
  • 1 month warranty
More About this Item
This large, lint-free cloth dissipates surface static while safely cleaning delicate surfaces ranging from film and lenses to view screens. Knitted from an ultra-fine mesh of non-toxic synthetic fiber and non-abrasive conductive filament. Since the cloth is not chemically treated or wet, it will not lose its effectiveness when exposed to air and may be reused many times. Measures 10''W x 18''L.