This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® Specialty SPE Column, Phenyl, 200 mg Sorbent, 3 mL; 50/Pk
Representative image only.
Cole-Parmer Item # RK-06477-37
Provide complementary selectivity to C18 stationary phases
  • Manufactured with phenyl solvent
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)200
  • Total Volume Capacity (mL)3
  • SorbentPhenyl
Key Features
  • Manufactured with phenyl solvent
More About this Item
Phenyl SPE columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. Columns are made of polypropylene and feature Luer-slip outlet.