This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® SPE Format Flash Chromatography Column, slip tip, NH2, 10 g/70 mL; 16/pk
Representative image only.
Cole-Parmer Item # RK-06474-90
Ideal for first stage clean-up columns post synthesis using a vacuum manifold
  • Use to separate primary, secondary, and tertiary amines
  • NH2 shows better peak shapes with polar and basic compound—ideal for carbohydrates
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (g)10
  • Column Particle Size (µm)50
  • Total Volume Capacity (mL)70
  • SorbentNH3
Key Features
  • Use to separate primary, secondary, and tertiary amines
  • NH2 shows better peak shapes with polar and basic compound—ideal for carbohydrates
More About this Item
Easily connect to flash chromatography system using the Luer-slip connection. Use columns on a rack or stand using gravity flow or with vacuum processing stations. All columns are made from polypropylene and come with polyethylene frits.