This item is now discontinued. Please browse similar items
Kinesis TELOS Flash Chromatography Column Holder for Biotage® Isolera® system
Representative image only.
Kinesis Item # RK-06475-99
Attach dry loading column directly to the main flash column
  • Quick-release guide design enables fast TELOS flash column installation
  • Luer connections ensure leak-free chromatography
  • Applications: Flash Chromatography Column Holder for Biotage® Isolera® system
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsFlash Chromatography Column Holder for Biotage® Isolera® system
Key Features
  • Quick-release guide design enables fast TELOS flash column installation
  • Luer connections ensure leak-free chromatography
  • Applications: Flash Chromatography Column Holder for Biotage® Isolera® system
More About this Item
Flash column holders accommodate any combinations of TELOS dry loading columns (4 to 40 g) and TELOS flash columns (4 to 330 g). Easily increase the capacity by installing two 120 g columns in series. Column guides with finger-tight fittings provide a quick release for fast and easy column installation.
Select from a universal stand-alone system or an add-on for Biotage® Isolera® system. The stand-alone system features a 9-7/16" x 8-11/16" (24 x 22 cm) base that can be placed next to or on top of the chromatography system. The column holder for Biotage Isolera system attaches directly to the side of the flash system with the supplied guides.