This item is now discontinued. Please browse similar items
Kinesis Round-Bottom Tubes, 13x100mm, glass; 1000/pk
Representative image only.
Kinesis Item # RK-98700-00
Great for tissue culture and bacteriological work
  • Constructed of borosilicate glass
  • Clear test tubes make viewing samples easy
  • Measures 13 mm x 100 mm
  • Designed with 13-415 thread type
SPECS & DESCRIPTION
Specifications & Description
  • Tube MaterialBorosilicate glass
  • Tube ColorClear
  • Tube Bottom ShapeRound
  • Tube Length (mm)100
  • Tube Outside Diameter (OD) (mm)13
  • Cap IncludedNo
  • DescriptionRound Bottom Tubes, 13x100mm, glass; 1000/pk
Key Features
  • Constructed of borosilicate glass
  • Clear test tubes make viewing samples easy
  • Measures 13 mm x 100 mm
  • Designed with 13-415 thread type
More About this Item
Manufactured of borosilicate glass, these test tubes are ideal for general bacteriological and tissue culture applications. Test tubes feature a threaded neck. Caps are not included.