Kinesis KX Microwave Vial w/ Crimp Cap, 10 to 20 mL, Silicone/PTFE, 20 mm; 50/pk

Representative image only.
Cole-Parmer Item # RK-98720-22
Decrease reaction times in organic synthesis
  • Resealable silicone/PTFE septa allow multiple reagent additions and reaction monitoring
  • Manufactured from contamination-free Type I borosilicate glass—will not taint mixtures

PRODUCT OPTIONS

$218.06CAD / Pkg of 50

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)20
  • Tube MaterialBorosilicate glass
  • Tube Bottom ShapeRounded
  • Tube ColorClear
  • Cap TypeCrimp
  • Cap Diameter (mm)20
  • Septum TypeSilicone/PTFE
  • Special FeaturesMax pressure of 290 psi (20 bar)

Key Features

  • Resealable silicone/PTFE septa allow multiple reagent additions and reaction monitoring
  • Manufactured from contamination-free Type I borosilicate glass—will not taint mixtures

More About this Item

Use these reaction vials in the microwave to speed up the reaction time in organic synthesis, especially when using palladium (Pd), platinum (Pt), or other metal catalysts. These vials allow the reactions to be performed at temperatures above the boiling point of the solvents used. Heavy-walled glass is highly resistant to both chemicals and thermal shock. Order stirrer bars to use with vials to achieve a homogeneous mixture, ensuring reactions are completed in the minimum time.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat