This item is now discontinued. Please browse similar items
Heraeus Noblelight Hollow Cathode Lamp, cesium, 37 mm, uncoded
Representative image only.
Kinesis Item # RK-11949-87
Achieve reliable atomic absorption results with high-performance lamps for your spectrometers
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCesium
  • Compatible WithUncoded
  • Diameter (in)1.5
  • Diameter (mm)37
  • Description Hollow Cathode Lamp, cesium, 37 mm, uncoded
Key Features
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
More About this Item
Heraeus Noblelight cesium (Cs) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamp’s lifetime to 5000 mA hours. Choose from coded lamp versions for Perkin Elmer®, Varian/Agilent, and Unicam systems.