Cole-Parmer EPA Screw Cap, 24mm, White Polypropylene, Open, Butyl Red/PTFE Grey Septa; 1000/pk
Representative image only.
Cole-Parmer Item # RK-98704-19
Meets EPA requirements for testing environmental contaminants
  • Septa meet the requirements of the US Environmental Protection Agency (EPA)
  • Caps are constructed of durable white polypropylene
  • Select caps with UltraBond® septum to avoid sample contamination
  • Designed with an open cap and butyl/PTFE septa for the enhanced chemical compatibility
PRODUCT OPTIONS
$858.99CAD / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeScrew Cap
  • Cap Diameter (mm)24
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Septum TypeButyl Red/PTFE Grey
  • SterileNo
  • Special FeaturesOpen Cap
  • DescriptionEPA Screw Cap, 24mm, White Polypropylene, Open, Butyl Red/PTFE Grey Septa; 1000/pk
Key Features
  • Septa meet the requirements of the US Environmental Protection Agency (EPA)
  • Caps are constructed of durable white polypropylene
  • Select caps with UltraBond® septum to avoid sample contamination
  • Designed with an open cap and butyl/PTFE septa for the enhanced chemical compatibility
More About this Item
These white polypropylene (PP) caps and EPA septa are free of substances that might influence sample analysis for testing of potentially harmful environmental contaminants in water or soil samples. Select from septa only, cap only, or cap and septa combination.