This item is now discontinued. Please browse similar items
Ken-a-vision 7200HD Digital Microscope/Camera; 5.0 megapixel, USB
Representative image only.
Ken-a-vision Mfr # 7200HD – Item # RK-03908-32
Use as a stand-alone microscope/viewer or attach to a microscope for image capture
Often Bought with...
SPECS & DESCRIPTION
Specifications & Description
  • Maximum Magnification50x
  • Resolution5 MP
  • OutputUSB 2.0
  • PowerUSB
  • DescriptionDigital Microscope/Camera; 5.0 megapixel, USB
Includes
software.
More About this Item
Adjustable gooseneck and swivel head allow easy positioning at any angle. USB output and included software allow you to capture images, make notations and highlight specific areas directly on the image, make measurements, create amalgamations, and more.
Model 03908-32 has a pre-attached USB cable for plug-and-play capability. Simply load the software and plug the USB cable into your PC or laptop and you're ready to go! Interchangeable lens system accommodates all C-mount lenses.