This item is now discontinued. Please browse similar items
Ken-a-vision 1401KEM Digital Microscope Camera, 10x adapter
Representative image only.
Ken-a-vision Mfr # 1401KEM – Item # RK-03908-22
Connect a camera to your microscope quickly and easily
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Resolution2.0 MP
  • Image Sensor1/3\" CMOS
  • OutputUSB 2.0
  • Cable(s)USB
  • Data Transfer30 frames/second
  • Height (InchShort)6.5
  • Depth (in)5.5
  • Width (InchShort)4.5
  • Height (cm)16.5
  • Depth (cm)14
  • Width (cm)11.4
  • PowerUSB
  • DescriptionDigital Microscope Camera, 10x adapter
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Capture high-resolution images (JPG, BMP, TIFF), movies, or time-lapsed sequences with this 2.07 megapixel microscope camera. Output your images to a PC or laptop through the USB port using the attached USB cable; power is supplied through the USB port.
Model 03908-22 features a microscope connection with a built-in 10x lens that takes the place of a stereomicroscope eyepiece; simply unscrew the existing eyepiece and replace with the camera.