This item is now discontinued. Please browse similar items
Kap-Uts 2007-25013 closures, 25 mm, natural, PP
Representative image only.
Kap-Uts Mfr # 2007-25013 – Item # RK-04501-09
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
SPECS & DESCRIPTION
Specifications & Description
  • Descriptionclosures, 25 mm, natural, PP
Key Features
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
More About this Item
These new tube closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.