This item is now discontinued. Please browse similar items
Kanomax Airflow Transducer without Display
Representative image only.
Kanomax Mfr # 6332 – Item # RK-10006-02
Analog output makes it easy to integrate with other equipment
  • Purchase probes separately and choose the ones that meet your application needs!
  • Smart probe technology lets you change probes without recalibrating the transducer
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)3
  • Height (in)1 1/8
  • Length (in)5
  • DescriptionAirflow Transducer without Display
  • Warranty1 year
1 Year Warranty
Key Features
  • Purchase probes separately and choose the ones that meet your application needs!
  • Smart probe technology lets you change probes without recalibrating the transducer
  • 1 year warranty
More About this Item
Airflow transducers are designed for fixed installation—great for production line quality control and monitoring local exhaust ventilation system. Transducers feature 0 to 5 VDC or 4 to 20 mA analog output so you can send your data to other instruments such as data loggers, alarms, or controllers. Choose from a unit with a display for on-site