Justrite PP12752 Quick-Disconnect Disposal Safety Can, Polyethylene, 2 Gal
Representative image only.
Justrite Mfr # PP12752 – Item # RK-86395-00
Remove can and empty waste without disconnecting inlet tubing
  • Automatically vents at 3-5 psig
  • Accepts 3/8" ID tubing
  • FM approved
  • OSHA and NFPA compliant
  • EPA complaint
  • 1 year warranty
PRODUCT OPTIONS
$659.71CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene(PE)
  • Capacity (Gal)2
  • Capacity (Liters)7.57
  • Exterior Height (in)14.75
  • Exterior Diameter (in)12
  • Exterior Height (cm)37.47
  • Exterior Diameter (cm)30.48
  • ApprovalFM
  • DescriptionQuick-Disconnect Disposal Cans, 2 gal
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Includes
Polypropylene fittings for 3/8" tubing.
Key Features
  • Automatically vents at 3-5 psig
  • Accepts 3/8" ID tubing
  • FM approved
  • OSHA and NFPA compliant
  • EPA complaint
  • 1 year warranty
More About this Item
Polyethylene disposal safety cans provide a safe, hands-free way to drain waste liquid from your HPLC applications. Protect the liquid and vapors inside from external heat sources. Quick-disconnect are valved open when connected and valved closed when disconnected for spill free changeout of safety cans without disconnecting the tubing.
Coalescing carbon filter attaches to one of the fittings to filter VOCs. Durable polyethylene is translucent for quick visual inspection of liquid level. Pouring spout includes stainless steel flame arrester to prevent flashback ignition. Dual fittings (intake, vent) eliminate back pressure and provide controlled handling of vapors.