Justrite 7225130 Steel Safety Can for Flammables, Type II, 1" Metal Hose, AccuFlow, 2.5 Gal

Representative image only.
Justrite Mfr # 7225130 – Item # RK-81791-45
Safely contain hazardous liquids and control vapors
  • A necessary part of compliance for safety in the workplace
  • Reduce risk of fire, protecting both personnel and facilities
  • Improve efficiency with ease-of-use features to enhance pouring and filling operations
  • FM approved
  • UL/ULC listed
  • TÜV certified
  • OSHA and NFPA compliant
  • 10 year warranty

PRODUCT OPTIONS

$299.83CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialSteel
  • Capacity (Gal)2.5
  • Capacity (Liters)9.46
  • Exterior Height (in)12
  • Exterior Diameter (in)11.75
  • Exterior Height (cm)30.48
  • Exterior Diameter (cm)29.86
  • ComplianceNFPA, OSHA
  • ApprovalFM, UL, cUL
  • DescriptionSafety Can for Flammables, Type II, 2.5 gallon
  • Warranty10 Years
10 Year WarrantyFM Approved

Key Features

  • A necessary part of compliance for safety in the workplace
  • Reduce risk of fire, protecting both personnel and facilities
  • Improve efficiency with ease-of-use features to enhance pouring and filling operations
  • FM approved
  • UL/ULC listed
  • TÜV certified
  • OSHA and NFPA compliant
  • 10 year warranty

More About this Item

All cans include a 3 1/2” built-in, stainless-steel flame arrester to prevent flashback ignition. Self-closing, spring loaded lid automatically vents at 3-5 psi (0.2 and 0.35 bar) to fight rupture or explosion. Reinforcing ribs resist denting or implosion. Steel construction with powder coat paint resists corrosion and improves wear and tear. A large ID zone allows you to label content of the can to avoid misuse

Type II safety cans feature patented AccuFlow™ manifold which is vented for smooth, fast, glug-free flow. Easy-to-use Safe-Squeeze® trigger provides controlled and precision pouring. The cans offer two openings: one for filling and one for pouring. Bottom lip provides grip and resists punctures.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat