Justrite 28179 HPLC Poly Manifold w/Stainless Steel Stacker Connector for up to 12 Waste Lines
Representative image only.
Justrite Mfr # 28179 – Item # RK-86394-31
A cost-effective way to collect HPLC waste
PRODUCT OPTIONS
$476.64CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene(PP)
  • DescriptionHPLC Poly Manifold w/stainless \"stacker\" connector
More About this Item
These polypropylene manifolds accept up to six waste lines from HPLC processes to be fed into a single intake fitting. Manifolds have six 1/4-28 ports, one 3/8" NPT port (all come with caps for unused ports), and one quick-disconnect port in either 316 stainless steel or polypropylene.
This manifold has a "stacker" connection for attaching on top of model 83694-27 or -29. Up to 12 waste lines can be attached to a single disposal can when using the stacker manifold.