John Guest PPSV011223W Speedfit Ball Valve, straight, 3/8" OD speedfit x 3/8" NPT(M)
Representative image only.
John Guest Mfr # PPSV011223W – Item # RK-07391-02
$24.61CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection Type3/8" OD Speedfit to 3/8" NPT(M)
  • Max Pressure (PSI)150
  • Max Pressure (bar)10.3
  • Max Temperature (° F)150
  • Max Temperature (° C)65
  • Wetted Materialsfood-grade polypropylene, EPDM, and stainless steel
  • DescriptionSpeedfit Ball Valve, straight, 3/8" OD speedfit x 3/8" NPT(M)
More About this Item
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.