Jenway 924 005 General Purpose, Glass-Bodied pH Electrode for Standard Measurements of General Solutions
Representative image only.
Jenway Item # RK-99960-35
A variety of pH electrodes for your Jewnay meters
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
PRODUCT OPTIONS
$162.07CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsStandard measurements of general solutions
  • Electrode TypeCombination
  • Electrode Body MaterialGlass
  • Reference JunctionAnnular ceramic
  • Reference ElectrolyteAgCl, liquid filled single junction
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1
  • DescriptionGeneral Purpose, Glass-Bodied pH Electrode for Standard Measurements of General Solutions
Key Features
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
More About this Item
Choose the right electrode for your application.