This item is now discontinued. Please browse similar items
Jenway 740301 Peltier Cell Holder
Representative image only.
Jenway Item # RK-83056-42
Optimize performance of your Jenway® spectrophotometer
  • Expand analysis capabilities or make testing easier
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithJenway 7410, 7415, and 7615 Spectrophotometers
  • DescriptionPeltier Cell Holder
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Expand analysis capabilities or make testing easier
  • 1 year warranty
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
For applications where sample temperature is critical, the Peltier accessory is offered which enables sample temperature to be set and controlled. The current temperature is displayed above the set point temperature on the LCD, and can be set in °C or °F.