This item is now discontinued. Please browse similar items
J. H. Williams Tool Group 7502MRMH Micro-Adjustable Click-Type Torque Wrench, 3/8" Drive, 100-750 in lb
Representative image only.
J. H. Williams Tool Group Mfr # 7502MRMHW – Item # RK-10780-54
Micro-Adjustable torque wrenches Metal Handle English Scale
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lb-in)750
  • Capacity (N-m)81.9
  • Drive (in)0.375
  • Length (in)16
  • Capacity750 in lb / 81.9 N m
  • DescriptionMicro-Adjustable Click-Type Torque Wrench, 3/8\" Drive, 100-750 in lb
More About this Item
The metal handle series offers the user a rugged, industrial strength torque wrench able to withstand the rigors of professional use. Handles are lightly knurled for a tough, non-slip grip. There are 34 models of metal handle micrometer adjustable torque wrenches available.
*Dual scale, calibrated dual direction
*Positive lock with spring loaded pull down lock ring
*Fast, accurate and easy to set
*Easy-to-read laser marked scales
*Most feature Quick Release buttons which provide good socket retention as well as easy socket removal
*Accuracy: +/- 4% clockwise +/- 6% CCW of indicated value, CW, from 20% to 100% of full scale
*Meets or exceeds ASME B107.14M-1994, ISO 6789 Standards
*Individually serialized with matching certificate of calibration traceable to N.I.S.T.