This item is now discontinued. Please browse similar items
J. H. Williams Tool Group 6004MFRMH Micro-Adjustable Click-Type Torque Wrench, 3/4" Drive, 100-600 ft lb
Representative image only.
J. H. Williams Tool Group Mfr # 6004MFRMH – Item # RK-10780-53
Micro-Adjustable torque wrenches Metal Handle English Scale
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (N-m)779
  • Capacity (lb-ft)600
  • Drive (in)0.75
  • Length (in)42
  • Capacity600 ft lb / 779 N m
  • DescriptionMicro-Adjustable Click-Type Torque Wrench, 3/4\" Drive, 100-600 ft lb
More About this Item
The metal handle series offers the user a rugged, industrial strength torque wrench able to withstand the rigors of professional use. Handles are lightly knurled for a tough, non-slip grip. There are 34 models of metal handle micrometer adjustable torque wrenches available.
*Dual scale, calibrated dual direction
*Positive lock with spring loaded pull down lock ring
*Fast, accurate and easy to set
*Easy-to-read laser marked scales
*Most feature Quick Release buttons which provide good socket retention as well as easy socket removal
*Accuracy: +/- 4% clockwise +/- 6% CCW of indicated value, CW, from 20% to 100% of full scale
*Meets or exceeds ASME B107.14M-1994, ISO 6789 Standards
*Individually serialized with matching certificate of calibration traceable to N.I.S.T.