This item is now discontinued. View the direct replacement: RK-18100-24 or browse similar items
International Products Corp M-1053 Micro Green Clean Biodegradable Cleaner, 215 KG (198 L)
Representative image only.
International Products Corp Mfr # M-1053 – Item # RK-18100-12
A free-rinsing, industrial strength, biodegradable hard surface cleaner designed for use in a wide range of applications.
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Dilution1:100 to 2:100, pH 9.8
  • ApplicationsGels, Emulsifiers, Oil, Titanium Doixides, Dyes, Silicaons, Fragrances, Petrolatum, Paraffins
  • Description M-1053 MICRO GREEN CLEAN BIODEGRADABLE CLEANER 215 NET WT PLASTIC DRUM
More About this Item
Excellent for cleaning metals, ceramics, medical instruments, food-processing equipment, filter membranes, and other surfaces. Typical pH: 9.5.