This item is now discontinued. We recommend the following as suggested replacements: RK-06231-05
Inoculating Loop, Yellow, 9" Long, 10 uL/1 mm ID Loop; 960/Box
Representative image only.
Item # RK-06231-31
Save time and eliminate cross-contamination
  • Sterile to eliminate cross-contamination
  • Color coded for easy identification
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Loop Inside Diameter (mm)4
  • Overall Length (in)9
  • Contained Volume (µL)10
  • Handle TypeLoop
  • ColorYellow
  • DescriptionInoculating Loop, Yellow, 9\" Long, 10 uL/1 mm ID Loop; 960/Box
Key Features
  • Sterile to eliminate cross-contamination
  • Color coded for easy identification
More About this Item
Prevent the risk of cross-contamination with these sterile, disposable needles and loops. Packaged in boxes of 960 (32 per bag, 30 bags per box).