InnoCal NIST-Traceable Calibration; pH and Conductivity Meter, Handheld/Benchtop
Representative image only.
InnoCal Item # RK-17106-20
Services that ensure the accuracy of your pH Measurement instrumentation
  • Five electrical pH and mV test points
PRODUCT OPTIONS
$242.37CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeLiquid Testing
  • Instrument Only CalibrationYes
  • Number Of Calibration Test Points5 Points
  • Calibration Test PointsAcross range
  • DescriptionNIST-Traceable Calibration, for a pH and Conductivity Meter
NIST Traceable Calibration
Key Features
  • Five electrical pH and mV test points
More About this Item
Regulatory and quality agencies like FDA and EPA have strict guidelines for the determination and control of pH. Maintain your compliance with the help of InnoCal. We can calibrate new instrument purchases from Cole-Parmer, recalibrate your existing instruments, and provide maintenance and repair services.
InnoCal’s metrologists can service most any brand of pH, conductivity, or dissolved oxygen meter including Oakton Instruments, accumet, Fisher Brand, Mettler-Toledo, YSI, WTW, Horiba, Hanna Instruments, Thermo Scientific Orion, VWR SympHony, and others.
Calibrations are performed on any handheld or benchtop pH meter with a BNC connection. The calibration contains five test points for pH (2, 4, 7, 10, 12) and five test points in millivolts across the range. If included, the automatic temperature compensation (ATC) probe will also be tested.