This item is now discontinued. Please browse similar items
Imada AP-003 Optional Test Stand Extender Plate
Representative image only.
Imada Mfr # AP-003 – Item # RK-93955-16
Expand the testing capabilities of your force gauge
  • Durable test stands are suitable for a wide range of applications
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)220
  • DescriptionOptional Test Stand Extender Plate
  • Warranty2 year
Two year warranty
Key Features
  • Durable test stands are suitable for a wide range of applications
  • 2 year warranty
More About this Item
Test stands produce more repeatable results than handheld testing. Several different sizes, loading methods, and capacities are available. All stands are ruggedly designed for many years of service in laboratory or production environments. Force gauge is sold separately.
Increases the standard test stand clearance to 10".