This item is now discontinued. Please browse similar items
Imada 905691 Data Acquisition Software Distance Meter Output Cable
Representative image only.
Imada Mfr # 905691 – Item # RK-93955-15
Capture key data from your torque testers
  • Customized Excel spreadsheet designed for capturing and analyzing peak and first peak data
  • Ideal for torque testers with either RS-232 or USB ports
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionData Acquisition Software Distance Meter Output Cable
  • Warranty2 year
Two year warranty
Key Features
  • Customized Excel spreadsheet designed for capturing and analyzing peak and first peak data
  • Ideal for torque testers with either RS-232 or USB ports
  • 2 year warranty
More About this Item
Gather peak torque or first peak readings as well as a running log of all torque measurements along with a chart of the data and statistical information. All data is entered directly into a standard Excel spreadsheet, allowing you to analyze your torque tester data using all of the powerful tools available in Excel.
Use with SW-2 data acquisition software and DB-2 data button to capture and display the current force and displacement values. Compatible with test stands equipped with distance meter.