This item is now discontinued. Please browse similar items
Idex V-540 Manual Injection Valve, Red, Polyimide/PTFE, 1/8" OD Tubing; 1/EA
Representative image only.
Idex Mfr # V-540 – Item # RK-02015-83
Introduce a sample of a solution into a flowing stream of liquid
  • 2-position, 6-port configuration
  • Red
  • 1/4-28 flat-bottom ports for 1/8" OD tubing
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4-28 flat bottom
  • Inlet Port1/4-28 flat bottom
  • Max Pressure (PSI)1000
  • Max Pressure (bar)69
  • Wetted MaterialsPolyimide/PTFE
  • Tubing OD (in)0.125
  • DescriptionManual Injection Valve, Red, Polyimide/PTFE, 1/8\" OD Tubing; 1/EA
  • Warranty1 year
1 Year Warranty
Includes
Six flangeless natural PEEK fitting with short head for 1/8" OD tubing 02020-99.
Key Features
  • 2-position, 6-port configuration
  • Red
  • 1/4-28 flat-bottom ports for 1/8" OD tubing
  • 1 year warranty
More About this Item
Manual injection valves are ideal for applications that require operator control or injection of smaller sample volumes. Injection valves are a type of switching valve usually featuring an attached sample loop (a length of tubing and fittings that match the angle of the valve ports).
Select from single- or dual-mode valves. Single-mode injection valves are configured for rear loading and requires complete filling of the sample loop. Dual-mode injection valves are configured for front loading and allows either partial or complete filling of the loop. Dual-mode valve introduces sample by syringe through the needle port built into the valve shaft. Valves with MBB design creates a continuous flow between the load and inject positions to eliminate pressure transient shock to the system.