This item is now discontinued. Please browse similar items
Idex P-445NF Micro-Metering Valve Without Fittings, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Mfr # P-445NF – Item # RK-02016-91
Excellent for fine control of fluid flow rates
  • Easily control fluid output for a wide range of low flow applications
  • 1/4-28 flat-bottom configuration
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Valve MaterialPEEK, PTFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 flat-bottom
  • Volume (µL)7.2
  • ColorBlack
  • DescriptionMicro-Metering Valve Without Fittings, 1/4-28 Flat-Bottom; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Easily control fluid output for a wide range of low flow applications
  • 1/4-28 flat-bottom configuration
  • 1 year warranty
More About this Item
Micro-metering valves can reduce outgoing flow to as low as 3.5 µL/min. Use valves in peristaltic pump fluid transfer applications, mass spectroscopy, fraction collection, and helium sparging lines. Valves have 0.020" (0.5 mm) thru holes.
Valve has 1/4-28 flat-bottom ports and has 0.020" (0.5 mm) thru holes. Provide your own fitting by matching the port configuration.
Volume listed is maximum volume with valve fully open.