This item is now discontinued. View the direct replacement: RK-12024-77 or browse similar items
Idex P-300 Flangeless Ferrule, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/Ea
Representative image only.
Idex Mfr # P-300 – Item # RK-02019-82
No need to flange tubing
  • Excellent for connecting rigid or semi-rigid tubing
  • Yellow ETFE
  • 1/EA
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorYellow
  • Warranty1 year
1 Year Warranty
Key Features
  • Excellent for connecting rigid or semi-rigid tubing
  • Yellow ETFE
  • 1/EA
  • 1 year warranty
More About this Item
Use with male nuts for a leak-tight connection.