This item is now discontinued. Please browse similar items
Idex P-208W Flangeless Male Nut, Standard Head, White Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Mfr # P-208W – Item # RK-02019-48
Short head is ideal for tight-space applications
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • White acetal
  • 1/EA
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorWhite
  • Warranty1 year
1 Year Warranty
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • White acetal
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.