This item is now discontinued. Please browse similar items
Idex 9000-1541 Debubbler, ETFE, No Degasser Tail; 5 mL
Representative image only.
Idex Mfr # 9000-1541 – Item # RK-42712-34
Eliminate interference from bubbles
  • Quickly, efficiently capture and remove dissolved gases
  • Reduce instrument downtime and improve performance
  • Integrate into any stand-alone degasser, degassing tray, or pump
  • Reduce partial reagent dispenses, creating fewer false positives
  • 5 mL maximum bubble capacity
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")6 51/64
  • Membrane MaterialPoridex
  • Housing MaterialETFE
  • Connections1/4-28
  • Max Pressure (PSI)30
  • Max Temperature (° C)40
  • DescriptionDebubbler, ETFE, No Degasser Tail; 5 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • Quickly, efficiently capture and remove dissolved gases
  • Reduce instrument downtime and improve performance
  • Integrate into any stand-alone degasser, degassing tray, or pump
  • Reduce partial reagent dispenses, creating fewer false positives
  • 5 mL maximum bubble capacity
  • 1 year warranty
More About this Item
Integrating a debubbler into the fluid path creates stable instrument performance across systems using applications that require dissolved gas by capturing and removing bubbles. You can choose components to actively remove bubbles with or without removing dissolved gases in the system. Debubblers reduce downtime and waste due to bubble formation and improve performance across your system.
Models that include a degasser remove bubbles from the liquid stream by vacuum through a Poridex™ membrane. Degassing is ideal for preventing bubble formation from recurring downstream.