This item is now discontinued. Please browse similar items
Idex 6000-209 316 Stainless Steel Fitting Assembly, 1/16" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex Mfr # 6000-209 – Item # RK-02022-28
Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar)!
  • Constructed of durable stainless steel
  • Standard length
  • 10-32 coned configuration
  • 10/PK
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless steel
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.588
  • Max Operating Pressure (PSI)20000
  • Description316 Stainless Steel Fitting Assembly, 1/16\" OD Tubing, 10-32 Coned; 10/PK
Includes
Ferrule 02022-21.
Key Features
  • Constructed of durable stainless steel
  • Standard length
  • 10-32 coned configuration
  • 10/PK
More About this Item
These 316 stainless steel fitting assemblies include nuts and ferrules are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
Metal fittings should not be used with plastic ports because they can damage the ports.